Learn More
Norgen Biotek Corp Cytoplasmic & Nuclear RNA Purification Kit, 50 Prep
Catalog No. NC1467197
Encompass_Preferred
This kit provides a rapid method for the isolation and purification of both cytoplasmic and nuclear RNA from cultured animal cells and small tissue samples. The kit can be used to isolate all sizes of RNA from the cytoplasmic and nuclear RNA fractions, including all small RNA species without any requirement for phenol. Included in the kit are sufficient reagents to perform either 50 cytoplasmic RNA preparations or 25 cytoplasmic and 25 nuclear RNA preparations. Ten samples can be processed in approximately 45 minutes. This kit is also available in a 100 prep size.
Catalog No. NC1467197 Supplier Norgen Biotek Corp Supplier No. 21000
This kit provides a rapid method for the isolation and purification of both cytoplasmic and nuclear RNA from cultured animal cells and small tissue samples. The kit can be used to isolate all sizes of RNA from the cytoplasmic and nuclear RNA fractions, including all small RNA species without any requirement for phenol. Included in the kit are sufficient reagents to perform either 50 cytoplasmic RNA preparations or 25 cytoplasmic and 25 nuclear RNA preparations. Ten samples can be processed in approximately 45 minutes. This kit is also available in a 100 prep size.