Learn More
Ethos Biosciences Eosin Y, Intensified, 1gal, Ethos
Catalog No. 5000574132
Encompass_Preferred
Eosin Y Intensified is a ready-to-use cytoplasmic counterstain formulated to provide enhanced staining intensity in routine histology and cytology applications. Designed for use in hematoxylin and eosin (H&E) procedures, it produces deeper pink to red coloration of cytoplasm, connective tissue, and extracellular components, improving contrast and tissue definition. The intensified formulation supports consistent color development and is suitable for manual or automated staining systems. Ideal for clinical, research, and educational laboratories, Eosin Y Intensified delivers reliable, reproducible results for routine tissue evaluation and morphology studies.
Catalog No. 50-005-74132 Supplier Ethos Biosciences Supplier No. 7007G
Eosin Y Intensified is a ready-to-use cytoplasmic counterstain formulated to provide enhanced staining intensity in routine histology and cytology applications. Designed for use in hematoxylin and eosin (H&E) procedures, it produces deeper pink to red coloration of cytoplasm, connective tissue, and extracellular components, improving contrast and tissue definition. The intensified formulation supports consistent color development and is suitable for manual or automated staining systems. Ideal for clinical, research, and educational laboratories, Eosin Y Intensified delivers reliable, reproducible results for routine tissue evaluation and morphology studies.