Learn More
Ethos Biosciences Eosin Y 1%, Alcoholic, 1gal, Ethos
Catalog No. 5000574131
Encompass_Preferred
Eosin Y 1% Alcoholic is a ready-to-use cytoplasmic counterstain formulated for routine histology and cytology applications. Commonly used in hematoxylin and eosin (H&E) staining procedures, it provides clear pink to red coloration of cytoplasmic components, connective tissue, and extracellular structures, enhancing contrast with nuclear stains. The alcoholic formulation supports rapid penetration and consistent staining intensity, making it suitable for manual and automated staining systems. Designed for use in clinical, research, and educational laboratories, Eosin Y 1% Alcoholic delivers reliable, reproducible results for routine tissue evaluation.
Catalog No. 50-005-74131 Supplier Ethos Biosciences Supplier No. 7006G
Eosin Y 1% Alcoholic is a ready-to-use cytoplasmic counterstain formulated for routine histology and cytology applications. Commonly used in hematoxylin and eosin (H&E) staining procedures, it provides clear pink to red coloration of cytoplasmic components, connective tissue, and extracellular structures, enhancing contrast with nuclear stains. The alcoholic formulation supports rapid penetration and consistent staining intensity, making it suitable for manual and automated staining systems. Designed for use in clinical, research, and educational laboratories, Eosin Y 1% Alcoholic delivers reliable, reproducible results for routine tissue evaluation.