Learn More
Med Vet International CytoPrep Fix & Dry, 5 Slides, J0334CP5, Each
Catalog No. 5000583006
Encompass_Preferred
The CytoPrep is a combination device that safely heat-fixes cytologies and quickly dries slides for rapid microscopy results. Heat-fixing cells onto slides, especially when preparing ear cytologies or drying a urine slide before staining, is the best way to ensure that cells adhere flat onto the glass and fully accept stains with minimum wash-off. The traditional method of heat-fixing slides with a bunsen burner or lighter is considered too dangerous by many clinics, because of safety concerns of an open-flame, and is also inconsistent because of poor temperature control. Open flames also create black soot on the slide which must be wiped off prior to staining. The CytoPrep eliminates these concerns with a controlled warm surface that can accommodate up to 5 slides for rapid heat-fixing in as little as 1 minute. After slides are heat-fixed then stained and rinsed with water, the slides need to dry before viewing under the microscope.
Catalog No. 50-005-83006 Supplier Med Vet International Supplier No. JOR334CP5
The CytoPrep is a combination device that safely heat-fixes cytologies and quickly dries slides for rapid microscopy results. Heat-fixing cells onto slides, especially when preparing ear cytologies or drying a urine slide before staining, is the best way to ensure that cells adhere flat onto the glass and fully accept stains with minimum wash-off. The traditional method of heat-fixing slides with a bunsen burner or lighter is considered too dangerous by many clinics, because of safety concerns of an open-flame, and is also inconsistent because of poor temperature control. Open flames also create black soot on the slide which must be wiped off prior to staining. The CytoPrep eliminates these concerns with a controlled warm surface that can accommodate up to 5 slides for rapid heat-fixing in as little as 1 minute. After slides are heat-fixed then stained and rinsed with water, the slides need to dry before viewing under the microscope.