Learn More
Electron Microscopy Sciences CytoSeal™ Mounting Medium 60 Mounting Medium 16 OZ
SDP
Catalog No. 5025272
Encompass_Preferred
CytoSeal 60 and 280 are toluene-based mounting mediumformulated from acrylic resins that will not crack or discolor with age. An antioxidant has been added to the formulation to inhibit fading or yellowing of stained specimens. It will not become brittle and crack, nor will it discolor or yellow with age. CytoSeal can be dissolved in toluene or xylene. Low-viscosity medium dries quickly and spreads evenly and rapidily virtually eliminating air bubbles.
Catalog No. 50-252-72 Supplier Electron Microscopy Sciences Supplier No. 18007
CytoSeal 60 and 280 are toluene-based mounting mediumformulated from acrylic resins that will not crack or discolor with age. An antioxidant has been added to the formulation to inhibit fading or yellowing of stained specimens. It will not become brittle and crack, nor will it discolor or yellow with age. CytoSeal can be dissolved in toluene or xylene. Low-viscosity medium dries quickly and spreads evenly and rapidily virtually eliminating air bubbles.