Learn More
Electron Microscopy Sciences CytoSeal 60, 4 oz
SDP
Catalog No. 5025273
Encompass_Preferred
Premium quality; formulated from the highest quality advanced acrylic resin. It will not become brittle and crack, nor will it discolor or yellow with age. CytoSeal dries rapidly, allowing examination soon after application. It can be used with oil immersion objectives and in fluorescent procedures. Slides will not stick together and the medium will not "cold flow" to the edge of the slide during long term storage. CytoSeal can be dissolved in toluene or xylene. An antioxidant has been added to the formulation to prevent fading of slides. CytoSeal comes in a Controlled Drop Dispenser, with varying viscosities. CytoSeal 60, which has a viscosity of 60cps, is intended primarily for use with cover glass.
Catalog No. 50-252-73 Supplier Electron Microscopy Sciences Supplier No. 18006
Premium quality; formulated from the highest quality advanced acrylic resin. It will not become brittle and crack, nor will it discolor or yellow with age. CytoSeal dries rapidly, allowing examination soon after application. It can be used with oil immersion objectives and in fluorescent procedures. Slides will not stick together and the medium will not "cold flow" to the edge of the slide during long term storage. CytoSeal can be dissolved in toluene or xylene. An antioxidant has been added to the formulation to prevent fading of slides. CytoSeal comes in a Controlled Drop Dispenser, with varying viscosities. CytoSeal 60, which has a viscosity of 60cps, is intended primarily for use with cover glass.