Learn More
Cell Biolabs Inc CytoSelect™ 24-Well Cell Invasion Assay Kit (Basement Membrane, Colorimetric), 12 assays
Catalog No. NC9993891
Encompass_Preferred
Cell invasion is the ability of malignant tumor cells to invade normal surrounding tissue, which contributes in large part to the significant morbidity and mortality of cancers. The CytoSelect™ Cell Invasion Assay Kit utilizes polycarbonate membrane inserts (8 µm pore size) to assay the invasive properties of cells. The kit contains twelve 8 um polycarbonate membrane inserts coated with an ECM gel layer in a 24-well plate. Cells must degrade the ECM layer and then move through the pores of the membrane. Invasive cells are then quantified using the provided Cell Stain Solution.
Catalog No. NC9993891 Supplier Cell Biolabs Inc Supplier No. CBA110
Cell invasion is the ability of malignant tumor cells to invade normal surrounding tissue, which contributes in large part to the significant morbidity and mortality of cancers. The CytoSelect™ Cell Invasion Assay Kit utilizes polycarbonate membrane inserts (8 µm pore size) to assay the invasive properties of cells. The kit contains twelve 8 um polycarbonate membrane inserts coated with an ECM gel layer in a 24-well plate. Cells must degrade the ECM layer and then move through the pores of the membrane. Invasive cells are then quantified using the provided Cell Stain Solution.