Learn More
Cell Biolabs Inc CytoSelect™ 24-Well Cell Invasion Assay Kit (Collagen I, Colorimetric), 12 assays
Catalog No. 50672641
Encompass_Preferred
Cell invasion is the ability of malignant tumor cells to invade normal surrounding tissue, which contributes in large part to the significant morbidity and mortality of cancers. The CytoSelect™ Cell Invasion Assay Kit utilizes polycarbonate membrane inserts (8 µm pore size) to assay the invasive properties of cells. The kit contains twelve 8 um polycarbonate membrane inserts coated with a Collagen gel layer in a 24-well plate. Cells must degrade the Collagen layer and then move through the pores of the membrane. Invasive cells are then quantified using the provided Cell Stain Solution.
Catalog No. 50-672-641 Supplier Cell Biolabs Inc Supplier No. CBA110COL
Add to Cart
Add to Cart
Cell invasion is the ability of malignant tumor cells to invade normal surrounding tissue, which contributes in large part to the significant morbidity and mortality of cancers. The CytoSelect™ Cell Invasion Assay Kit utilizes polycarbonate membrane inserts (8 µm pore size) to assay the invasive properties of cells. The kit contains twelve 8 um polycarbonate membrane inserts coated with a Collagen gel layer in a 24-well plate. Cells must degrade the Collagen layer and then move through the pores of the membrane. Invasive cells are then quantified using the provided Cell Stain Solution.