Learn More
Cell Biolabs Inc CytoSelect™ 96-Well Cell Invasion Assay Kit (Basement Membrane, Fluorometric), 96 assays
Supplier:  Cell Biolabs Inc CBA112
Encompass_Preferred
Cell invasion is the ability of malignant tumor cells to invade normal surrounding tissue, which contributes in large part to the significant morbidity and mortality of cancers. The CytoSelect™ Cell Invasion Assay Kit utilizes polycarbonate membrane inserts (8 um pore size) to assay the invasive properties of cells. The kit contains 96 polycarbonate membrane inserts with 8 um pores coated with an ECM gel layer in a 96-well plate. Cells must degrade the ECM layer and then move through the pores of the membrane. Invasive cells are then quantified using the provided fluorometric dye.
Catalog No. NC9382145