Learn More
Cell Biolabs Inc CYTOSELECT WOUND HEALING 24W
Catalog No. NC9849005
Encompass_Preferred
The CytoSelect 24-Well Wound Healing Assay provides more consistent results well-to-well compared to homemade scratch assays. Useful for measuring cell migration, cell proliferation and wound closure.
Catalog No. NC9849005 Supplier Cell Biolabs Inc Supplier No. CBA120
Add to Cart
Add to Cart
The CytoSelect 24-Well Wound Healing Assay provides more consistent results well-to-well compared to homemade scratch assays. Useful for measuring cell migration, cell proliferation and wound closure.