Learn More
Cell Biolabs Inc CytoSelect™ WST-1 Cell Proliferation Assay Reagent, 960 assays
Catalog No. NC0713545
Encompass_Preferred
The CytoSelect™ WST-1 Cell Proliferation Assay Reagent provides a colorimetric format for measuring and monitoring cell proliferation. Cells can be plated and then treated with compounds or agents that affect proliferation. Cells are then detected with the proliferation reagent, which is converted in live cells from WST-1 to the formazan form in the presence of cellular NADH and an electron mediator. An increase in cell proliferation is accompanied by increased signal, while a decrease in cell proliferation (and signal) can indicate the toxic effects of compounds or suboptimal culture conditions. The assay principles are basic and can be applied to most eukaryotic cell lines, including adherent and non-adherent cells and certain tissues. This reagent can be used to detect proliferation in bacteria, yeast, fungi, protozoa as well as cultured mammalian and piscine cells. The reagent is sufficient for the evaluation of 960 assays in 96-well plates or 192 assays in 24-well plates.
Catalog No. NC0713545 Supplier Cell Biolabs Inc Supplier No. CBA253
The CytoSelect™ WST-1 Cell Proliferation Assay Reagent provides a colorimetric format for measuring and monitoring cell proliferation. Cells can be plated and then treated with compounds or agents that affect proliferation. Cells are then detected with the proliferation reagent, which is converted in live cells from WST-1 to the formazan form in the presence of cellular NADH and an electron mediator. An increase in cell proliferation is accompanied by increased signal, while a decrease in cell proliferation (and signal) can indicate the toxic effects of compounds or suboptimal culture conditions. The assay principles are basic and can be applied to most eukaryotic cell lines, including adherent and non-adherent cells and certain tissues. This reagent can be used to detect proliferation in bacteria, yeast, fungi, protozoa as well as cultured mammalian and piscine cells. The reagent is sufficient for the evaluation of 960 assays in 96-well plates or 192 assays in 24-well plates.