Learn More
Advanced Biomatrix CytoSilk Lyophilized Silk Fibroin 1 gram
Catalog No. 502255944
Encompass_Preferred
This Silk Fibroin lyophilized powder is approximately 1 gram of lyophilized protein derived from the Bombyx Mori silkworm. Silk Fibroin can be reconstituted at various concentrations and is useful for forming various scaffolds such as coatings, films, sponges, hydrogels and electrospun fibers.
This item is not returnable. View return policy
Catalog No. 50-225-5944 Supplier Advanced Biomatrix Supplier No. 53521GM
This Silk Fibroin lyophilized powder is approximately 1 gram of lyophilized protein derived from the Bombyx Mori silkworm. Silk Fibroin can be reconstituted at various concentrations and is useful for forming various scaffolds such as coatings, films, sponges, hydrogels and electrospun fibers.