Learn More
Cayman Chemical Cytosine-13C,15N2, Purity: ≥98%, CAS: 181517-10-2, 10 mg
SDP
Catalog No. 502945263
Encompass_Preferred
An internal standard for the quantification of cytosine by GC- or LC-MS
Catalog No. 50-294-5263 Supplier Cayman Chemical Supplier No. 4112210 MG
An internal standard for the quantification of cytosine by GC- or LC-MS