Learn More
Genscript Corporation CytoSinct 1000 Cell Isolation
Supplier:  Genscript Corporation D00023
Encompass
GenScript has developed the CytoSinct 1000 Cell Isolation system for easy automated isolation or depletion of human cells Scientists can now isolate up to 120x10 9 total cells (full leukopak) in a single run within 2 hours
Catalog No. 50-000-57087
Add to Cart
Request a Quote