Learn More
Genscript Corporation CytoSinct CD34 Nanobeads hum
Catalog No. 5000057100
Encompass
The CytoSinct CD34 Nanobeads human are used for in vitro enrichment CD34 cells from fresh or frozen peripheral blood mononuclear cells (PBMCs) leukapheresis products or single cell suspension based on the surface expression of human CD34
Catalog No. 50-000-57100 Supplier Genscript Corporation Supplier No. L009671
The CytoSinct CD34 Nanobeads human are used for in vitro enrichment CD34 cells from fresh or frozen peripheral blood mononuclear cells (PBMCs) leukapheresis products or single cell suspension based on the surface expression of human CD34