Learn More
Genscript Corporation CytoSinct CD34 Nanobeads hum
Catalog No. 5000058628
Encompass
CytoSinct CD34 Nanobeads human (GMP) is intended for the in vitro magnetic labeling for pre-separation or other processing of human CD34 positive cells from heterogeneous cell populations enabling further cultivation and/or genetic modification of the cells
Catalog No. 50-000-58628 Supplier Genscript Corporation Supplier No. L00948
CytoSinct CD34 Nanobeads human (GMP) is intended for the in vitro magnetic labeling for pre-separation or other processing of human CD34 positive cells from heterogeneous cell populations enabling further cultivation and/or genetic modification of the cells