Learn More
Genscript Corporation CytoSinct CD4 Nanobeads huma
Supplier:  Genscript Corporation L008631
Encompass
CytoSinct CD4 Nanobeads human is used for in vitro enrichment of CD4 cells from fresh or frozen peripheral blood mononuclear cells (PBMCs) leukapheresis products or single-cell suspension based on the surface expression of human CD4
Catalog No. 50-000-58414