Learn More
Genscript Corporation CytoSinct CD4 Nanobeads huma
Catalog No. 5000059126
Encompass
CytoSinct CD4 Nanobeads human is used for in vitro enrichment of CD4 cells from fresh or frozen peripheral blood mononuclear cells (PBMCs) leukapheresis products or single-cell suspension based on the surface expression of human CD4
Catalog No. 50-000-59126 Supplier Genscript Corporation Supplier No. L008630.5
CytoSinct CD4 Nanobeads human is used for in vitro enrichment of CD4 cells from fresh or frozen peripheral blood mononuclear cells (PBMCs) leukapheresis products or single-cell suspension based on the surface expression of human CD4