Learn More
Genscript Corporation CytoSinct CD4 Nanobeads huma
Catalog No. 5000057394
Encompass
The CytoSinct CD4 Nanobeads human (GMP) is used for in vitro isolation or depletion CD4 cells from fresh or frozen peripheral blood mononuclear cells (PBMCs) Ieukapheresis products or single cell suspension based on the surface expression of human CD4 Due to the nanoscale structure of the CytoSinct CD4 Nanobeads human (GMP) the excess reagent can be removed by centrifugation or conventional supernatant replacement
Catalog No. 50-000-57394 Supplier Genscript Corporation Supplier No. L009327.5
The CytoSinct CD4 Nanobeads human (GMP) is used for in vitro isolation or depletion CD4 cells from fresh or frozen peripheral blood mononuclear cells (PBMCs) Ieukapheresis products or single cell suspension based on the surface expression of human CD4 Due to the nanoscale structure of the CytoSinct CD4 Nanobeads human (GMP) the excess reagent can be removed by centrifugation or conventional supernatant replacement