Learn More
Genscript Corporation CytoSinct CD4 Nanobeads, human, immunomagnetic, colloidal solution, anti-human CD4 antibody, iron-dextran composition, 2-8 °C storage, non-toxic, GMP compliant
Catalog No. NC3715831
Encompass
CytoSinct CD4 Nanobeads, human, immunomagnetic, colloidal solution, anti-human CD4 antibody, iron-dextran composition, 2-8 °C storage, non-toxic, GMP compliant
Catalog No. NC3715831 Supplier Genscript Corporation Supplier No. L00932
CytoSinct CD4 Nanobeads, human, immunomagnetic, colloidal solution, anti-human CD4 antibody, iron-dextran composition, 2-8 °C storage, non-toxic, GMP compliant