Learn More
Genscript Corporation CytoSinct CD8 Nanobeads, GMP, Volume 2.5 mL, Bead Composition Iron-dextran conjugated to anti-human CD8 antibody, Storage Temperature 2-8 °C
Catalog No. NC3715832
Encompass
CytoSinct CD8 Nanobeads, GMP, Volume 2.5 mL, Bead Composition Iron-dextran conjugated to anti-human CD8 antibody, Storage Temperature 2-8 °C
Catalog No. NC3715832 Supplier Genscript Corporation Supplier No. L00933
CytoSinct CD8 Nanobeads, GMP, Volume 2.5 mL, Bead Composition Iron-dextran conjugated to anti-human CD8 antibody, Storage Temperature 2-8 °C