Learn More
Genscript Corporation CytoSinct gL Columns
Catalog No. 5000057090
Encompass
GenScript s CytoSinct gL Columns have been developed for gentle isolation of cells labeled with superparamagnetic CytoSinct Nanoparticles Due to the extremely small size of the CytoSinct Nanobeads high strength magnetic field is required to retain the labeled cells The CytoSinct gL Columns contain an optimized matrix to generate a strong magnetic field when placed in a permanent magnet such as the CytoSinct Magnet
Catalog No. 50-000-57090 Supplier Genscript Corporation Supplier No. D00008
GenScript s CytoSinct gL Columns have been developed for gentle isolation of cells labeled with superparamagnetic CytoSinct Nanoparticles Due to the extremely small size of the CytoSinct Nanobeads high strength magnetic field is required to retain the labeled cells The CytoSinct gL Columns contain an optimized matrix to generate a strong magnetic field when placed in a permanent magnet such as the CytoSinct Magnet