Learn More
Genscript Corporation CytoSinct gM Columns
Catalog No. 5000058896
Encompass
GenScript s CytoSinct gM Columns have been developed for gentle isolation of cells labeled with superparamagnetic CytoSinct Nanoparticles Due to the extremely small size of the CytoSinct Nanobeads high strength magnetic field is required to retain the labeled cells The CytoSinct gM Columns contain an optimized matrix to generate a strong magnetic field when placed in a permanent magnet such as the CytoSinct Magnet
Catalog No. 50-000-58896 Supplier Genscript Corporation Supplier No. D00007
GenScript s CytoSinct gM Columns have been developed for gentle isolation of cells labeled with superparamagnetic CytoSinct Nanoparticles Due to the extremely small size of the CytoSinct Nanobeads high strength magnetic field is required to retain the labeled cells The CytoSinct gM Columns contain an optimized matrix to generate a strong magnetic field when placed in a permanent magnet such as the CytoSinct Magnet