Learn More
Genscript Corporation CytoSinct L1 magnet
Supplier:  Genscript Corporation D00011
Encompass
The CytoSinct Manual Separator system is developed to separate human and animal cells The material to be separated for example cells is first magnetically labeled with superparamagnetic CytoSinct Nanobeads After magnetic labeling cells are passed through a CytoSinct Columns which is placed in the strong permanent magnet of the CytoSinct Manual Separator The ferromagnetic spheres in the column amplify the magnetic field by 10 000-fold thus inducing a high Magnetic field gradient Unlabeled cells pass through while magnetically labeled cell are retained within the column After removal of the column from the magnetic field the retained fraction can be eluted Both fractions labeled and non-labeled can be collected for downstream applications including cell culture and flow cytometry
Catalog No. 50-000-58958