Learn More
Sigma Aldrich Fine Chemicals Biosciences Cytosine beta-D-arabinofuranoside, 147-94-4, MFCD00066487, 100 mg
Catalog No. NC0707739
Encompass
Empirical Formula (Hill Notation): C9H13N3O5, Molecular Weight: 243.22, crystalline, ≥90% (HPLC), storage temp: 2-8°C, Synonym: (β-D-Arabinofuranosyl)cytosine, Ara-C, Arabinocytidine, Arabinosylcytosine, Cytarabine, Cytosine arabinoside.
Catalog No. NC0707739 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C1768100MG
Empirical Formula (Hill Notation): C9H13N3O5, Molecular Weight: 243.22, crystalline, ≥90% (HPLC), storage temp: 2-8°C, Synonym: (β-D-Arabinofuranosyl)cytosine, Ara-C, Arabinocytidine, Arabinosylcytosine, Cytarabine, Cytosine arabinoside.