Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosine beta-D-arabinofuranoside hydrochloride, 99%, crystalline, Thermo Scientific Chemicals

Catalog No. AC227180010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227180010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227180010 Supplier Thermo Scientific Chemicals Supplier No. 227180010
  • $333.00
    $262.65 / Each of 1

    Save $70.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 69-74-9 | Molecular Formula: C9H14ClN3O5 | Molecular Weight: 279.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-74-9
Molecular Formula C9H14ClN3O5
Molecular Weight (g/mol) 279.68
MDL Number MFCD00012839
InChI Key KCURWTAZOZXKSJ-JBMRGDGGSA-N
Synonym cytarabine hydrochloride
PubChem CID 134129637
SMILES Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Melting Point 197.0°C to 198.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 10, 2778
Specific Rotation +127° to +133° (20°C, 589nm) ((c=0.7, H2O))
IUPAC Name 4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
Formula Weight 279.67
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Cytosine β-D-arabinofuranoside hydrochloride
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Germ cell mutagenicity (category 2), Reproductive toxicity (category 2), Skin sensitisation (category 1)
Hazard Statement Causes serious eye irritation, Suspected of causing genetic defects, Suspected of damaging the unborn child, May cause an allergic skin reaction
EINECSNumber 200-713-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.