Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosine-β-D-Arabinofuranoside, Hydrochloride, MP Biomedicals™

Catalog No. ICN10007101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10007101 1 g
ICN10007105 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10007101 Supplier MP Biomedicals, Inc Supplier No. 0210007101
Add to Cart
Add to Cart

An anti-leukemic agent resulting in DNA fragmentation and inhibition of replication

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-74-9
Molecular Formula C9H14ClN3O5
Molecular Weight (g/mol) 279.68
MDL Number MFCD00012839
InChI Key KCURWTAZOZXKSJ-JBMRGDGGSA-N
Synonym cytarabine hydrochloride
PubChem CID 134129637
IUPAC Name 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride
SMILES Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Color White
Quantity 1 g
Formula Weight 279.7
Physical Form Powder
Chemical Name or Material Cytosine-β-D-Arabinofuranoside, Hydrochloride
EINECSNumber 200-713-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.