Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytosine-d2 | 106391-24-6 | MFCD01317453 | 99.7% | 113.11 g/mol | C4H3D2N3O | 10 MG
SDP

Catalog No. 5000297684
Encompass_Preferred

Cytosine-d2 is a deuterium-labeled isotopic standard of the nucleobase cytosine intended for analytical and biochemical applications, including mass spectrometry, tracer experiments, and method development. It is supplied as a dry solid and requires appropriate handling and storage.

  • Deuterium-labeled cytosine (d2) for isotope dilution and tracer studies.
  • Suitable as an internal standard for LC-MS and GC-MS quantification.
  • High purity, approximately 99.7%.
  • Soluble in DMSO at 10 mg/mL with ultrasonic and warming; hygroscopic DMSO may affect solubility.
  • Store at 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-97684 Supplier Medchemexpress LLC Supplier No. HYI0626S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytosine-d2 is a deuterium-labeled isotopic standard of the nucleobase cytosine intended for analytical and biochemical applications, including mass spectrometry, tracer experiments, and method development. It is supplied as a dry solid and requires appropriate handling and storage.

  • Deuterium-labeled cytosine (d2) for isotope dilution and tracer studies.
  • Suitable as an internal standard for LC-MS and GC-MS quantification.
  • High purity, approximately 99.7%.
  • Soluble in DMSO at 10 mg/mL with ultrasonic and warming; hygroscopic DMSO may affect solubility.
  • Store at 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.