Learn More
Abcam Cytosine, Nucleoside, 10G
Catalog No. 502704792
Encompass_Preferred
MW 111.1 g/mol. Nucleoside. Pyrimidine derivative. One of the four main bases found in DNA and RNA, along with adenine, guanine, and thymine (uracil in RNA). Cytosine reagent is used in a wide variety of research applications, as an enzyme substrate or precursor of effector molecules such as cytosine sugars.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4792 Supplier Abcam Supplier No. AB14625710G
MW 111.1 g/mol. Nucleoside. Pyrimidine derivative. One of the four main bases found in DNA and RNA, along with adenine, guanine, and thymine (uracil in RNA). Cytosine reagent is used in a wide variety of research applications, as an enzyme substrate or precursor of effector molecules such as cytosine sugars.
The product is subject to the following: Abcam Restricted Use Statement