Learn More
Biotium Cytoskeletal Stains Cell and Organelle Stains Cytoskeletal Stains CF488 1/EA
Catalog No. 5000499674
Encompass_Preferred
Vitamin D-Binding Protein conjugated to green fluorescent CF488A can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF488A has an Ex/Em of 490/516 nm
Catalog No. 50-004-99674 Supplier Biotium Supplier No. 70087
Vitamin D-Binding Protein conjugated to green fluorescent CF488A can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF488A has an Ex/Em of 490/516 nm