Learn More
Biotium Cytoskeletal Stains Cell and Organelle Stains Cytoskeletal Stains CF594 1/EA
Catalog No. 5000499709
Encompass_Preferred
Vitamin D-Binding Protein conjugated to red fluorescent CF594 can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF594 has an Ex/Em of 593/615 nm
Catalog No. 50-004-99709 Supplier Biotium Supplier No. 70088
Vitamin D-Binding Protein conjugated to red fluorescent CF594 can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF594 has an Ex/Em of 593/615 nm