Learn More
Biotium Cytoskeletal Stains Cell and Organelle Stains Cytoskeletal Stains CF640 1/EA
Catalog No. 5000499427
Encompass_Preferred
Vitamin D-Binding Protein conjugated to far-red fluorescent CF640R can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF640R has an Ex/Em of 642/663 nm
Catalog No. 50-004-99427 Supplier Biotium Supplier No. 70089
Vitamin D-Binding Protein conjugated to far-red fluorescent CF640R can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF640R has an Ex/Em of 642/663 nm