Learn More
Biotium Cytoskeletal Stains Cell and Organelle Stains Cytoskeletal Stains CF647 1/EA
Catalog No. 5000499764
Encompass_Preferred
Vitamin D-Binding Protein conjugated to far-red fluorescent CF647 can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF647 has an Ex/Em of 652/668 nm
Catalog No. 50-004-99764 Supplier Biotium Supplier No. 70090
Vitamin D-Binding Protein conjugated to far-red fluorescent CF647 can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF647 has an Ex/Em of 652/668 nm