Learn More
Biotium Cytoskeletal Stains Cell and Organelle Stains Cytoskeletal Stains CF740 1/EA
Catalog No. 5000499843
Encompass_Preferred
Vitamin D-Binding Protein conjugated to near-infrared fluorescent CF740 can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF740 has an Ex/Em of 742/767 nm
Catalog No. 50-004-99843 Supplier Biotium Supplier No. 70091
Vitamin D-Binding Protein conjugated to near-infrared fluorescent CF740 can be used to stain monomeric G-actin in fixed and permeabilized cells The conjugate has been optimized for immunofluoresnce and is is supplied at 100 ug/mL CF740 has an Ex/Em of 742/767 nm