Learn More
Epredia™ Cytoslide™ Microscope Slides
Catalog No. 5991051 Shop All Epredia Products
Change view
Click to view available options
Description:
Coated, circle on back
Noncoated, circle on front
Shandon™ Single Cytoslides™; Noncoated, circle on back
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
5991051 Shandon™ Single Cytoslides™; Noncoated, circle on back
12-070-266 Coated, circle on back
59-910-59 Noncoated, circle on front
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 5991051 Supplier Epredia™ Supplier No. 5991051
Add to Cart
Add to Cart
This item is not returnable. View return policy
Improve processing and visualization of small-volume specimens with these cytoslides, for use with Epredia™ Shandon™ EZ Single Cytofunnels™.
  • Uncoated slides available with circle on front or back
  • Available in cases of 100 slides
Specifications
Type Single Cytoslide
Color Plain
Description Shandon™ Single Cytoslides™; Noncoated, circle on back