Learn More
Cayman Chemical Cytosine-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 106391-24-6, 10 mg
SDP
Catalog No. 502912960
Encompass_Preferred
An internal standard for the quantification of cytosine by GC- or LC-MS
Catalog No. 50-291-2960 Supplier Cayman Chemical Supplier No. 3631910 MG
An internal standard for the quantification of cytosine by GC- or LC-MS