Learn More
Cayman Chemical Cytosine-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 106391-24-6, 25 mg
SDP
Catalog No. 502913013
Encompass_Preferred
An internal standard for the quantification of cytosine by GC- or LC-MS
Catalog No. 50-291-3013 Supplier Cayman Chemical Supplier No. 3631925 MG
An internal standard for the quantification of cytosine by GC- or LC-MS