Learn More
Advanced Biomatrix CytoSoft Imaging 24-well Plate, Elastic Modulus 2 kPa
Catalog No. NC1578604
Encompass_Preferred
This CytoSoft Imaging product, Catalog Number 5185, has an elastic modulus of approximately 2 kPa with a polycarbonate film-bottom, black flat-bottom 24-well plate. The thickness of the silicone gel is uniform with a ~0.03 mm thick layer of silicone in each well. The rigidity of the substrate to which cells adhere can have a profound effect on cell morphology and gene expression. The CytoSoft Imaging plate is for high-resolution imaging where low autofluorescence and exceptional optical clarity are required. Plate consists of a #1.5 glass bottom plate bonded with a black polystyrene frame and includes a lid. The plates are sterilized using ozone irradiation and provided with 1 plate per package. On the bottom of each well, there is a thin layer of specially formulated biocompatible silicone, whose elastic modulus (rigidity) is carefully measured and certified.
Catalog No. NC1578604 Supplier Advanced Biomatrix Supplier No. 51851EA
This CytoSoft Imaging product, Catalog Number 5185, has an elastic modulus of approximately 2 kPa with a polycarbonate film-bottom, black flat-bottom 24-well plate. The thickness of the silicone gel is uniform with a ~0.03 mm thick layer of silicone in each well. The rigidity of the substrate to which cells adhere can have a profound effect on cell morphology and gene expression. The CytoSoft Imaging plate is for high-resolution imaging where low autofluorescence and exceptional optical clarity are required. Plate consists of a #1.5 glass bottom plate bonded with a black polystyrene frame and includes a lid. The plates are sterilized using ozone irradiation and provided with 1 plate per package. On the bottom of each well, there is a thin layer of specially formulated biocompatible silicone, whose elastic modulus (rigidity) is carefully measured and certified.