Learn More
Epredia™ Cytospin™ 4 Cytocentrifuge Sealed Head Rotor
GSA_VA
Catalog No. 59910019 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Head Seals
Sealed Head Rotor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
59910019 Head Seals
59910018 Sealed Head Rotor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 59910019 Supplier Epredia™ Supplier No. 59910019
Add to Cart
Add to Cart
Use these accessories with the Epredia™ Cytospin™ 4 Cytocentrifuge.
  • Shorten preparation time by having a second sealed head ready when the first finishes processing
  • Autoclavable
  • 12-chamber head seals completely
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) Cytospin 4 Cytocentrifuge
Product Type Head Seals