Learn More
Cayman Chemical Cytosporone B, Purity: ≥95%, CAS: 321661-62-5, 10 mg
SDP
Catalog No. 502965847
Encompass_Preferred
A naturally occurring agonist for the nuclear receptor Nur77 (IC50 = 0.278 nM); induces apoptosis in cancer cells and retards xenograft tumor growth; induces genes related to gluconeogenesis and decreases atherosclerosis progression in mice fed a high fat/high cholesterol diet
Catalog No. 50-296-5847 Supplier Cayman Chemical Supplier No. 1750110 MG
A naturally occurring agonist for the nuclear receptor Nur77 (IC50 = 0.278 nM); induces apoptosis in cancer cells and retards xenograft tumor growth; induces genes related to gluconeogenesis and decreases atherosclerosis progression in mice fed a high fat/high cholesterol diet