Learn More
Medchemexpress LLC HY-N2148 2mg Medchemexpress, Cytosporone B CAS:321661-62-5 Purity:>98%
Supplier: Medchemexpress LLC HYN21482MG
Medchemexpress, HY-N2148 2mg Cytosporone B CAS:321661-62-5 Cytosporone B (Csn-B; Dothiorelone G) is a naturally occurring nuclear orphan receptor Nur77/NR4A1 agonist with an EC 50 of 0.278 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.