Learn More
Andwin Scientific CYTOTEK 1ML BASE HOLDER 200/CS
SDP
Catalog No. 50727372
Encompass_Preferred
Cytotek 1mL Base Holder is an equipment that goes along with Cytotek Centrifuge Funnel Chamber that helps enhance cell recovery with the use of Cytotek Cytocentrifuge. Sold in 200 units per case.
Catalog No. 50-727-372 Supplier Andwin Scientific Supplier No. 6261055
Cytotek 1mL Base Holder is an equipment that goes along with Cytotek Centrifuge Funnel Chamber that helps enhance cell recovery with the use of Cytotek Cytocentrifuge. Sold in 200 units per case.