Learn More
Andwin Scientific CYTOTEK 1ML CHAMBER CAP 200/CS
SDP
Catalog No. 50727373
Encompass_Preferred
The Cyto-Tek chamber cap is an accessory for the Cyto-Tek Centrifuge Funnel Chamber, a product made to reduce cell attachment and improve cell recovery up to 100%. The chamber cap has a volume capacity of 1 mL and is sold in a case of 200. The funnel chamber and its other accessories are all sold separately.
Catalog No. 50-727-373 Supplier Andwin Scientific Supplier No. 6261060
The Cyto-Tek chamber cap is an accessory for the Cyto-Tek Centrifuge Funnel Chamber, a product made to reduce cell attachment and improve cell recovery up to 100%. The chamber cap has a volume capacity of 1 mL and is sold in a case of 200. The funnel chamber and its other accessories are all sold separately.