Learn More
Andwin Scientific CYTOTEK SPECIMEN CHAMBER 200/B
SDP
Catalog No. NC9514438
Encompass_Preferred
The Cyto-Tek fluid chamber is an accessory for the Cyto-Tek Centrifuge Funnel Chamber, a product made to reduce cell attachment and improve cell recovery up to 100%. The base holder has a volume capacity of 1 mL and is sold in a case of 200. The funnel chamber and its other accessories are all sold separately.
Catalog No. NC9514438 Supplier Andwin Scientific Supplier No. 4329
The Cyto-Tek fluid chamber is an accessory for the Cyto-Tek Centrifuge Funnel Chamber, a product made to reduce cell attachment and improve cell recovery up to 100%. The base holder has a volume capacity of 1 mL and is sold in a case of 200. The funnel chamber and its other accessories are all sold separately.