Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytotoxic DNA minor groove interstrand crosslinking pyrrolobenzodiazepine dimer | 1595275-71-0 | MFCD34469130 | 98.9% | 584.66 g/mol | C33H36N4O6 | 5 MG
SDP

Catalog No. 5000212773
Encompass_Preferred

SG3199 is a pyrrolobenzodiazepine (PBD) dimer that forms covalent DNA interstrand cross-links in the minor groove, producing potent cytotoxicity. It is used as the released warhead component of tesirine-based ADC payloads and as a research reagent for preclinical ADC and cytotoxicity studies. Supplied as a high-purity material with recommended low-temperature, light-protected storage.

  • Potent DNA minor-groove interstrand crosslinker.
  • Designed for ADC payload and preclinical cytotoxicity research.
  • High purity (98.9%).
  • Molecular weight 584.66 g/mol; formula C33H36N4O6.
  • Storage: -20°C, protect from light, store under inert gas; in solvent: -80°C for extended storage.

Catalog No. 50-002-12773 Supplier Medchemexpress LLC Supplier No. HY1011615MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SG3199 is a pyrrolobenzodiazepine (PBD) dimer that forms covalent DNA interstrand cross-links in the minor groove, producing potent cytotoxicity. It is used as the released warhead component of tesirine-based ADC payloads and as a research reagent for preclinical ADC and cytotoxicity studies. Supplied as a high-purity material with recommended low-temperature, light-protected storage.

  • Potent DNA minor-groove interstrand crosslinker.
  • Designed for ADC payload and preclinical cytotoxicity research.
  • High purity (98.9%).
  • Molecular weight 584.66 g/mol; formula C33H36N4O6.
  • Storage: -20°C, protect from light, store under inert gas; in solvent: -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.