Learn More
Sigma Aldrich Fine Chemicals Biosciences Cytotoxicity Detection Kit
Catalog No. 502905780
Encompass
A nonradioactive alternative to the [51Cr] and [3H]-thymidine release assay.Colorimetric assay for the quantification of cell death and cell lysis based on the measurement of lactate dehydrogenase (LDH) activity released from the cytosol of damaged cells into the supernatant.
Catalog No. 50-290-5780 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11644793001
A nonradioactive alternative to the [51Cr] and [3H]-thymidine release assay.Colorimetric assay for the quantification of cell death and cell lysis based on the measurement of lactate dehydrogenase (LDH) activity released from the cytosol of damaged cells into the supernatant.