Learn More
Cytotoxicity detection Kit, MilliporeSigma™ Supelco™
Catalog No. 111004032 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 Tests
Catalog No. Quantity
11-100-4032 100 Tests
1 options
Catalog No. 11-100-4032 Supplier MilliporeSigma Supelco Supplier No. 919631KT
Add to Cart
Add to Cart
Colorimetric assay system. Utilize for both homo- and non-homogeneous assays. Can be used with and without transferring supernatant. Stable Working Solution (6 months at 0°C)
Cytotoxicity LDH Assay Kit-WST is a kit for determination of cytotoxicity by measuring a lactate dehydrogenase (LDH) activity released from damaged cells. LDH is a stable cytoplasmic enzyme present in all types of cells and released into the cell culture medium through damaged plasma membrane. Since Cytotoxicity LDH Assay kit-WST neither reflects the activity of living cells nor is harmful to cells, cytotoxicity can be measured with living cells (homogeneous assay). In addition, it is possible to perform a non-homogeneous assay by using the cell culture supernatant.After preparation the Working Solution is stable under refrigerated conditions and can readily be used for up to six months.
TRUSTED_SUSTAINABILITY
Specifications
Type Organic Standard
Quantity 100 Tests