Learn More
Dojindo Molecular Technologies Inc Cytotoxicity LDH Assay Kit-WST (500 TESTS) measures released LDH from dead cells and can be used with and without transferring supernatant.
SDP
Catalog No. NC1406882
Encompass
Lactate dehydrogenase(LDH) is an enzyme that is present in almost all cell types and it catalyzes the oxidation of lactate to pyruvate in the presence of co-enzyme NAD+. Once cells are impaired by stress, injuries, chemicals or intercellular signals, LDH is rapidly released from the cell membrane. Thus, the measurement of the amount of released LDH from cells is one of the major methods to assess cell death. Since Dojindo's Cytotoxicity LDH Assay Kit-WST neither reflects the activity of living cells nor is harmful to cells, it allows the assay to perform in wells containing both viable and damaged cells. Cell Cytotoxicity is evaluated by multiple cell functions. Dojindo provides you useful selection guide. (https://www.dojindo.com/cell-proliferation-cytotoxicity-assay-kits-selection-guide/)
Catalog No. NC1406882 Supplier Dojindo Molecular Technologies Inc Supplier No. CK1205
Lactate dehydrogenase(LDH) is an enzyme that is present in almost all cell types and it catalyzes the oxidation of lactate to pyruvate in the presence of co-enzyme NAD+. Once cells are impaired by stress, injuries, chemicals or intercellular signals, LDH is rapidly released from the cell membrane. Thus, the measurement of the amount of released LDH from cells is one of the major methods to assess cell death. Since Dojindo's Cytotoxicity LDH Assay Kit-WST neither reflects the activity of living cells nor is harmful to cells, it allows the assay to perform in wells containing both viable and damaged cells. Cell Cytotoxicity is evaluated by multiple cell functions. Dojindo provides you useful selection guide. (https://www.dojindo.com/cell-proliferation-cytotoxicity-assay-kits-selection-guide/)