Learn More
Axion Biosystems CYTOVIEW MEA 48-BLACK PLATE
SDP
Catalog No. NC1882732
Encompass
CytoView MEA 48-Black plate, 16 microelectrodes per well, black polystyrene walls with a transparent SU-8 bottom, box of 5 plates
Catalog No. NC1882732 Supplier Axion Biosystems Supplier No. M768TMEA48B5
CytoView MEA 48-Black plate, 16 microelectrodes per well, black polystyrene walls with a transparent SU-8 bottom, box of 5 plates