Learn More
Axion Biosystems Cytoview Microelectrode Array 96, Black 5/pk
SDP
Catalog No. NC1675973
Encompass
96 wells, 8 PEDOT electrodes/well, Transparent bottom. CytoView microelectrode array (MEA) plates for the Maestro Pro, Maestro Edge, and Maestro Apex MEA systems combine unparalleled access to cellular electrical network information with a thin, transparent plate bottom for culture visualization and assay multiplexing. CytoView MEA plates contain the industry-leading electrode count per well, deliver low-noise signals, and can be read over days, weeks, or months.
Catalog No. NC1675973 Supplier Axion Biosystems Supplier No. M768TMEA96B5
96 wells, 8 PEDOT electrodes/well, Transparent bottom. CytoView microelectrode array (MEA) plates for the Maestro Pro, Maestro Edge, and Maestro Apex MEA systems combine unparalleled access to cellular electrical network information with a thin, transparent plate bottom for culture visualization and assay multiplexing. CytoView MEA plates contain the industry-leading electrode count per well, deliver low-noise signals, and can be read over days, weeks, or months.